Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173469.1 | NP_001166940.1 | 1 | 850 | ||
GenBank mRNA | BC169092.1 | - | 1 | 850 | 5599968 |