Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105712.1 | NP_001099182.1 | 4 | 2092 | ||
XM_008762681.1 | XP_008760903.1 | 3 | 3785 | |||
XM_008762682.1 | XP_008760904.1 | 4 | 2568 | |||
XM_008762683.1 | XP_008760905.1 | 3 | 3785 | |||
GenBank mRNA | M92076.1 | - | 3 | 2010 |
Set Membership: |
![]() |