Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Atp5j2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271117.1 | NP_001258046.1 | 4 | 369 | ||
GenBank mRNA | FQ210117.1 | - | 4 | 356 | ||
FQ211282.1 | - | 4 | 356 | |||
FQ216873.1 | - | 4 | 344 | |||
FQ217338.1 | - | 4 | 343 | |||
FQ217676.1 | - | 4 | 344 | |||
FQ218321.1 | - | 4 | 358 | |||
FQ221351.1 | - | 4 | 348 | |||
FQ221691.1 | - | 4 | 356 | |||
FQ221732.1 | - | 4 | 338 | |||
FQ222492.1 | - | 4 | 343 | |||
FQ223220.1 | - | 4 | 343 | |||
FQ223295.1 | - | 4 | 346 | |||
FQ223575.1 | - | 4 | 341 | |||
FQ223738.1 | - | 4 | 344 | |||
FQ223886.1 | - | 3 | 322 | |||
FQ223939.1 | - | 4 | 344 | |||
FQ224192.1 | - | 4 | 344 | |||
FQ224234.1 | - | 4 | 358 | |||
FQ224423.1 | - | 4 | 356 | |||
FQ224479.1 | - | 4 | 356 | |||
FQ224557.1 | - | 4 | 356 | |||
FQ224602.1 | - | 4 | 357 | |||
FQ224807.1 | - | 4 | 356 | |||
FQ228438.1 | - | 4 | 344 |