Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044253.1 | NP_001037718.1 | 4, 5 | 967 | ||
XM_006243271.2 | XP_006243333.1 | 4, 5 | 1904 | |||
XM_008766324.1 | XP_008764546.1 | 5, 6 | 1623 | |||
XM_008766325.1 | XP_008764547.1 | 5, 6 | 1595 | |||
XM_008766326.1 | XP_008764548.1 | 5, 6 | 1554 | |||
XM_008766327.1 | XP_008764549.1 | 5, 6 | 1526 | |||
GenBank mRNA | DQ489562.1 | - | 4, 5 | 967 |