Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NR1I3 SIRNA Assays ›
Gene Symbol:
NR1I3
Gene Name:
nuclear receptor subfamily 1 group I member 3
Gene Aliases:
CAR, CAR1, MB67
Chromosome Location:
Chr.1: 161229666 - 161238623 on Build GRCh38
UniGene ID:
Hs.349642
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s19368
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NR1I3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001077469.2
NP_001070937.1
3
359
NM_001077470.2
NP_001070938.1
2
219
NM_001077471.2
NP_001070939.1
3
359
NM_001077472.2
NP_001070940.1
2
219
NM_001077473.2
NP_001070941.1
2
219
NM_001077474.2
NP_001070942.1
3
359
NM_001077475.2
NP_001070943.1
2
219
NM_001077476.2
NP_001070944.1
2
219
NM_001077477.2
NP_001070945.1
2
219
NM_001077478.2
NP_001070946.1
3
359
NM_001077479.2
NP_001070947.1
2
219
NM_001077480.2
NP_001070948.1
3
359
NM_001077481.2
NP_001070949.1
3
359
NM_001077482.2
NP_001070950.1
3
359
NM_005122.4
NP_005113.1
3
359
XM_005245693.4
XP_005245750.1
2
632
XM_005245694.4
XP_005245751.1
2
631
XM_005245697.4
XP_005245754.1
3
242
XM_011510237.2
XP_011508539.1
2
629
GenBank mRNA
AB307702.1
-
2
156
AK303757.1
-
2
212
AK303796.1
-
3
393
AK310773.1
-
2
100
AK316208.1
-
3
351
AY572806.1
-
3
250
AY572807.1
-
3
179
AY572808.1
-
2
110
AY572809.1
-
2
110
AY572810.1
-
3
250
AY572811.1
-
2
110
AY572812.1
-
2
110
AY572813.1
-
3
250
AY572814.1
-
2
110
AY572815.1
-
2
110
AY572816.1
-
2
110
AY572817.1
-
2
110
AY572818.1
-
2
110
AY572819.1
-
3
250
AY572820.1
-
2
110
AY572822.1
-
3
250
AY572823.1
-
3
250
AY572824.1
-
2
110
AY572825.1
-
2
110
AY572826.1
-
3
250
AY572827.1
-
2
110
BC069626.1
-
3
312
7262456
BC121120.1
-
3
260
40120664
BC121121.1
-
3
260
40120665
CB124867.1
-
3
359
DQ022681.1
-
2
156
HQ692838.1
-
2
156
HQ692839.1
-
2
156
HQ692840.1
-
2
156
HQ692841.1
-
2
156
HQ709178.1
-
2
156
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
osteoblast differentiation
transcription initiation from RNA polymerase II promoter
signal transduction
androgen receptor signaling pathway
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
androgen receptor activity
thyroid hormone receptor activity
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-6lbnp:80/10.239.161.162:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline