Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048205.1 | NP_001041670.1 | 8 | 1224 | ||
NM_005132.2 | NP_005123.2 | 9 | 1123 | |||
XM_011537421.1 | XP_011535723.1 | 8 | 1396 | |||
XM_017021841.1 | XP_016877330.1 | 8 | 1193 | |||
GenBank mRNA | AF006264.1 | - | 8 | 696 | ||
AK001435.1 | - | 9 | 1123 | |||
AK291191.1 | - | 9 | 1255 | |||
AL832973.1 | - | 8 | 1114 | |||
BC004159.2 | - | 8 | 780 | 2822680 | ||
BC010887.1 | - | 8 | 1157 | 3535425 | ||
BC019326.1 | - | 2 | 142 | |||
CR457263.1 | - | 8 | 696 |