Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DMTF1 SIRNA Assays ›
Gene Symbol:
DMTF1
Gene Name:
cyclin D binding myb like transcription factor 1
Gene Aliases:
DMP1, DMTF, MRUL, hDMP1
Chromosome Location:
Chr.7: 87152361 - 87196332 on Build GRCh38
UniGene ID:
Hs.196129
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s19406
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DMTF1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142326.1
NP_001135798.1
15
1890
NM_001142327.1
NP_001135799.1
16
2007
NM_021145.3
NP_066968.3
18
2244
NR_024549.1
16
2218
NR_024550.1
16
2179
XM_011516735.2
XP_011515037.1
16
2929
XM_011516737.1
XP_011515039.1
17
2054
XM_011516739.2
XP_011515041.1
15
1819
XM_017012865.1
XP_016868354.1
17
3023
XM_017012866.1
XP_016868355.1
15
2806
XM_017012867.1
XP_016868356.1
14
3474
XM_017012868.1
XP_016868357.1
17
2054
XM_017012869.1
XP_016868358.1
18
2083
XM_017012870.1
XP_016868359.1
15
1822
XM_017012871.1
XP_016868360.1
17
3195
XM_017012872.1
XP_016868361.1
16
3101
XM_017012873.1
XP_016868362.1
17
2990
XM_017012874.1
XP_016868363.1
17
2226
XM_017012875.1
XP_016868364.1
17
3029
XM_017012876.1
XP_016868365.1
15
1969
XM_017012877.1
XP_016868366.1
16
2088
XM_017012878.1
XP_016868367.1
18
3123
GenBank mRNA
AB209230.1
-
15
2066
AF052102.1
-
2
99
AF084530.1
-
16
1956
AF202144.1
-
16
2128
AF202145.1
-
16
2167
AK002073.1
-
4
466
AK123259.1
-
2
908
AK126664.1
-
16
2674
AK296211.1
-
15
1783
AK314622.1
-
16
1943
BC007418.2
-
7
956
BC007447.2
-
7
956
BC070064.1
-
18
2190
4373991
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
cell cycle
cell differentiation
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II transcription factor recruiting
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding
transcription regulatory region DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-j4kkz:80/10.239.163.216:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0