Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173533.1 | NP_001167004.1 | 8 | 1372 | ||
NM_001173534.1 | NP_001167005.1 | 8 | 1363 | |||
NM_001184781.1 | NP_001171710.1 | 9 | 1486 | |||
NM_005137.2 | NP_005128.1 | 9 | 1495 | |||
NR_033674.1 | 8 | 1380 | ||||
GenBank mRNA | AK291670.1 | - | 9 | 1453 | ||
AK295697.1 | - | 8 | 1338 | |||
AK296337.1 | - | 8 | 1321 | |||
AK303130.1 | - | 9 | 1405 | |||
AK304382.1 | - | 8 | 1330 | |||
BC020339.1 | - | 9 | 1442 | |||
BC032430.1 | - | 9 | 1431 | 5243412 | ||
BC035425.1 | - | 9 | 1442 | |||
BC040500.1 | - | 9 | 1490 | 5272271 | ||
CR456433.1 | - | 9 | 1267 | |||
CR936871.1 | - | 10 | 1830 | |||
D79985.1 | - | 9 | 1432 | |||
L46352.1 | - | 1 | 63 | |||
L77560.1 | - | 9 | 1396 | |||
X83545.1 | - | 9 | 1396 | |||
X84076.1 | - | 9 | 1393 |