Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000041.3 | NP_000032.1 | 4 | 1067 | ||
NM_001302688.1 | NP_001289617.1 | 4 | 1149 | |||
NM_001302689.1 | NP_001289618.1 | 4 | 998 | |||
NM_001302690.1 | NP_001289619.1 | 4 | 1098 | |||
NM_001302691.1 | NP_001289620.1 | 4 | 1082 | |||
GenBank mRNA | AL037046.3 | - | 1 | 222 | ||
BC003557.1 | - | 4 | 996 | 3355712 | ||
K00396.1 | - | 4 | 1011 | |||
M12529.1 | - | 4 | 1012 |