Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000714.5 | NP_000705.2 | 4 | 573 | ||
NM_001256530.1 | NP_001243459.1 | 4 | 768 | |||
NM_001256531.1 | NP_001243460.1 | 4 | 677 | |||
NR_046308.1 | 3 | 362 | ||||
GenBank mRNA | AF075589.1 | - | 3 | 376 | ||
AF075590.1 | - | 3 | 376 | |||
AY998017.1 | - | 3 | 468 | |||
BC001110.2 | - | 4 | 523 | 2989070 | ||
BG749252.1 | - | 4 | 677 | |||
BG752086.1 | - | 4 | 574 | |||
BQ057964.1 | - | 558 | ||||
BT006949.1 | - | 3 | 453 | |||
CR456409.1 | - | 3 | 462 | |||
CR457007.1 | - | 2 | 172 | |||
L21950.1 | - | 3 | 332 | |||
M36035.1 | - | 4 | 514 |