Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EVX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304519.1 | NP_001291448.1 | 3 | 1199 | ||
NM_001304520.1 | NP_001291449.1 | 4 | 1251 | |||
NM_001989.4 | NP_001980.1 | 3 | 1254 | |||
GenBank mRNA | AK298822.1 | - | 4 | 428 | ||
AK314155.1 | - | 3 | 1044 | |||
X60655.1 | - | 3 | 996 |