Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000858.5 | NP_000849.1 | 8, 9 | 858 | ||
NM_001159390.1 | NP_001152862.1 | 7, 8 | 693 | |||
NM_001159391.1 | NP_001152863.1 | 7, 8 | 607 | |||
NM_001242839.1 | NP_001229768.1 | 7, 8 | 633 | |||
XM_005273104.4 | XP_005273161.1 | 9, 10 | 1027 | |||
XM_017001092.1 | XP_016856581.1 | 8, 9 | 1346 | |||
XM_017001095.1 | XP_016856584.1 | 8, 9 | 803 | |||
XM_017001096.1 | XP_016856585.1 | 7, 8 | 643 | |||
GenBank mRNA | AK303845.1 | - | 7, 8 | 646 | ||
BC006249.2 | - | 8, 9 | 796 | 3946293 | ||
BC007369.2 | - | 6, 7 | 510 | |||
BC009914.2 | - | 8, 9 | 770 | 2821563 | ||
BC091480.1 | - | 7, 8 | 613 | 30340102 | ||
BE780558.1 | - | 7, 8 | 580 | |||
BI771824.1 | - | 7, 8 | 633 | |||
CR456836.1 | - | 6, 7 | 555 | |||
CR542006.1 | - | 6, 7 | 555 | |||
L76200.1 | - | 6, 7 | 555 | |||
U66895.1 | - | 6, 7 | 561 |