Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024844.1 | NP_001020015.1 | 9 | 1201 | ||
NM_002231.3 | NP_002222.1 | 10 | 1276 | |||
XM_006718223.2 | XP_006718286.1 | 11 | 1315 | |||
XM_011520067.2 | XP_011518369.1 | 7 | 978 | |||
GenBank mRNA | AK225410.1 | - | 10 | 1202 | ||
AK225708.1 | - | 10 | 1230 | |||
BC000726.2 | - | 10 | 1174 | 2959683 | ||
BC001821.2 | - | 10 | 1174 | 2959683 | ||
S48196.1 | - | 10 | 1194 | |||
U20770.1 | - | 10 | 1193 |