Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031804.2 | NP_001026974.1 | 1 | 6258 | ||
NM_005360.4 | NP_005351.2 | 2 | 2027 | |||
GenBank mRNA | AF055377.1 | - | 2 | 2011 | ||
AF447709.1 | - | 1 | 83 | |||
BC081542.1 | - | 1 | 5551 | 30347784 |