- Order Status
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: MPST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013436.2 | NP_001013454.1 | 2 | 449 | ||
NM_001130517.2 | NP_001123989.1 | 3 | 625 | |||
NM_021126.5 | NP_066949.2 | 2 | 439 | |||
XM_005261610.3 | XP_005261667.1 | 3 | 570 | |||
XM_011530195.1 | XP_011528497.1 | 2 | 392 | |||
XM_011530196.1 | XP_011528498.1 | 1 | 316 | |||
GenBank mRNA | AB208824.1 | - | 3 | 567 | ||
AK055733.1 | - | 2 | 1207 | |||
BC003508.1 | - | 1 | 322 | 3611253 | ||
BC009450.1 | - | 1 | 322 | |||
BC016737.1 | - | 2 | 412 | 4105509 | ||
BC018717.1 | - | 1 | 319 | 4131927 | ||
BI597227.1 | - | 2 | 445 | |||
BT019636.1 | - | 1 | 292 | |||
CR456523.1 | - | 1 | 311 | |||
CR541712.1 | - | 1 | 292 | |||
CX866643.1 | - | 2 | 332 | |||
CX872464.1 | - | 3 | 566 | |||
X59434.1 | - | 1 | 323 |