Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MT1M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176870.2 | NP_789846.1 | 3 | 408 | ||
GenBank mRNA | AF136177.1 | - | 3 | 385 | ||
BC028280.1 | - | 3 | 370 | 5191481 |
Gene Symbol: MT1JP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036677.1 | 3 | 377 | |||
GenBank mRNA | AF348994.2 | - | 3 | 377 |