Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRRG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000951.2 | NP_000942.1 | 3 | 302 | ||
NM_001316335.1 | NP_001303264.1 | 3 | 469 | |||
XM_006723286.2 | XP_006723349.1 | 4 | 305 | |||
XM_011527124.1 | XP_011525426.1 | 2 | 297 | |||
GenBank mRNA | AF009243.1 | - | 2 | 146 | ||
BC026032.1 | - | 2 | 496 | 3960215 | ||
CD013954.1 | - | 3 | 449 | |||
CD609900.1 | - | 3 | 310 | |||
CR456846.1 | - | 2 | 137 |