Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TPM3 SIRNA Assays ›
Gene Symbol:
TPM3
Gene Name:
tropomyosin 3
Gene Aliases:
CAPM1, CFTD, HEL-189, HEL-S-82p, NEM1, OK/SW-cl.5, TM-5, TM3, TM30, TM30nm, TM5, TPMsk3, TRK, hscp30
Chromosome Location:
Chr.1: 154155304 - 154192135 on Build GRCh38
UniGene ID:
Hs.535581
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s194839
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TPM3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001043351.1
NP_001036816.1
6
701
NM_001043352.1
NP_001036817.1
6
701
NM_001043353.1
NP_001036818.1
6
701
NM_001278188.1
NP_001265117.1
5
567
NM_001278189.1
NP_001265118.1
6
701
NM_001278191.1
NP_001265120.1
6
547
NM_152263.3
NP_689476.2
7
799
NM_153649.3
NP_705935.1
6
701
NR_103461.1
6
701
GenBank mRNA
AB062125.1
-
2
114
AF474157.1
-
6
571
AK026559.1
-
6
641
AK056889.1
-
2
690
AK056921.1
-
6
558
AK056997.1
-
7
589
AK092690.1
-
7
639
AK092712.1
-
5
446
AK123571.1
-
1
1347
AK125367.1
-
3
2585
AK298678.1
-
5
545
AK301674.1
-
6
523
AK308007.1
-
5
476
AK315504.1
-
6
660
AL537561.3
-
6
611
AY004867.1
-
6
582
BC000771.2
-
6
613
3506357
BC008407.1
-
7
765
4245271
BC008425.1
-
7
768
4246114
BC015403.1
-
6
616
4389911
BC062740.1
-
7
631
4271142
BC072428.1
-
6
628
6472532
BF967161.1
-
6
515
BG706843.1
-
6
644
BI546300.1
-
6
612
BU956481.1
-
6
627
BX419658.2
-
6
613
BX648485.1
-
6
613
CR997359.1
-
6
592
EU668324.1
-
7
639
FJ224335.1
-
6
613
KU178368.1
-
6
571
KU178369.1
-
6
571
KU178370.1
-
6
571
M12128.1
-
1
199
X04201.1
-
7
740
X04588.1
-
6
623
Back To Top
More Information
Panther Classification:
Molecular Function -
actin binding motor protein
Gene Ontology Categories:
Function(s)
Process(es)
movement of cell or subcellular component
muscle contraction
muscle filament sliding
molecular_function
actin binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-qqtbl:80/10.239.164.61:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0