Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004327.3 | NP_004318.3 | 14 | 3369 | ||
NM_021574.2 | NP_067585.2 | 14 | 3369 | |||
GenBank mRNA | AB209991.1 | - | 14 | 3118 | ||
AF192533.1 | - | 3 | 191 | |||
AF321981.1 | - | 2 | 134 | |||
AK303241.1 | - | 14 | 1803 | |||
BC066122.1 | - | 14 | 3200 | |||
BQ422888.1 | - | 5 | 495 | |||
X02596.1 | - | 14 | 3261 | |||
Y00661.1 | - | 14 | 3369 |