Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFITM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003641.3 | NP_003632.3 | 2 | 524 | ||
GenBank mRNA | AK290480.1 | - | 2 | 499 | ||
BC000897.1 | - | 2 | 474 | 3464598 | ||
BT007173.1 | - | 2 | 344 | |||
J04164.1 | - | 2 | 660 | |||
X84958.1 | - | 2 | 454 |