Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003857.3 | NP_003848.1 | 2 | 791 | ||
XM_011525427.2 | XP_011523729.1 | 4 | 1948 | |||
GenBank mRNA | AF040630.1 | - | 2 | 820 | ||
AF080586.1 | - | 2 | 733 | |||
BC069130.1 | - | 2 | 789 | 7216986 | ||
BC074914.2 | - | 2 | 755 | 30915339 | ||
BC074915.2 | - | 2 | 755 | 30915558 | ||
BC109051.1 | - | 2 | 758 | 40031879 | ||
BC109052.1 | - | 2 | 757 | 40031882 | ||
EF577401.1 | - | 2 | 708 |
Set Membership: |
G-protein coupled receptor |