Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302959.1 | NP_001289888.1 | 3 | 477 | ||
NM_001302960.1 | NP_001289889.1 | 3 | 511 | |||
NM_003977.3 | NP_003968.3 | 3 | 511 | |||
GenBank mRNA | BC104797.1 | - | 3 | 413 | 8143800 | |
BC104827.1 | - | 3 | 413 | 8143830 | ||
BG821308.1 | - | 3 | 506 | |||
BQ054206.1 | - | 3 | 477 | |||
BQ420398.1 | - | 2 | 237 | |||
U31913.1 | - | 3 | 390 | |||
U78521.1 | - | 3 | 498 |