Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR7E24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079935.1 | NP_001073404.1 | 1 | 63 | ||
GenBank mRNA | BC146920.1 | - | 1 | 177 | 9056848 |