Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERTAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013376.3 | NP_037508.2 | 2 | 888 | ||
GenBank mRNA | AF117959.1 | - | 2 | 859 | ||
AF366402.1 | - | 2 | 840 | |||
AK074652.1 | - | 2 | 888 | |||
AK222970.1 | - | 2 | 855 | |||
AK223011.1 | - | 2 | 855 | |||
BC002670.2 | - | 2 | 837 | 3613213 |