Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFRSF12A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016639.2 | NP_057723.1 | 4 | 1003 | ||
GenBank mRNA | AB035480.1 | - | 4 | 954 | ||
AB035481.1 | - | 3 | 849 | |||
AF191148.1 | - | 4 | 946 | |||
BC002718.2 | - | 4 | 974 | 3632233 |