Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GHRL SIRNA Assays ›
Gene Symbol:
GHRL
Gene Name:
ghrelin/obestatin prepropeptide
Gene Aliases:
MTLRP
Chromosome Location:
Chr.3: 10285750 - 10292947 on Build GRCh38
UniGene ID:
Hs.590080
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s195232
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GHRL
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001134941.2
NP_001128413.1
6
1228
NM_001134944.1
NP_001128416.1
4
358
NM_001134945.1
NP_001128417.1
4
355
NM_001134946.1
NP_001128418.1
3
241
NM_001302821.1
NP_001289750.1
7
982
NM_001302822.1
NP_001289751.1
6
770
NM_001302823.1
NP_001289752.1
6
767
NM_001302824.1
NP_001289753.1
6
692
NM_001302825.1
NP_001289754.1
5
495
NM_016362.4
NP_057446.1
6
1231
NR_126505.1
2
132
XM_017006612.1
XP_016862101.1
5
1766
XM_017006613.1
XP_016862102.1
5
1772
GenBank mRNA
AB029434.1
-
4
393
AB035700.1
-
4
379
AJ252278.1
-
4
417
AY184207.1
-
3
284
AY359053.1
-
5
535
BC025791.1
-
4
399
5212768
BM854032.1
-
5
409
EF139854.1
-
5
673
EF139855.1
-
5
470
EF139856.1
-
4
480
EF549557.1
-
2
102
EF549569.1
-
5
947
EF549571.1
-
6
740
EF549572.1
-
6
662
EF549573.1
-
5
465
EF549574.1
-
4
328
EF549575.1
-
3
211
EU072083.1
-
6
1201
EU072084.1
-
6
1198
EU072085.1
-
6
737
EU072086.1
-
4
325
EU072087.1
-
7
952
Back To Top
More Information
Panther Classification:
Molecular Function -
peptide hormone
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
gastric acid secretion
negative regulation of endothelial cell proliferation
glucose metabolic process
G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
actin polymerization or depolymerization
adult feeding behavior
response to hormone
hormone-mediated signaling pathway
dendrite development
negative regulation of angiogenesis
growth hormone secretion
positive regulation of insulin secretion
regulation of response to food
positive regulation of appetite
negative regulation of interleukin-1 beta production
negative regulation of locomotion
positive regulation of multicellular organism growth
regulation of cell proliferation
negative regulation of circadian sleep/wake cycle, REM sleep
negative regulation of tumor necrosis factor biosynthetic process
negative regulation of apoptotic process
cortisol secretion
response to estrogen
negative regulation of interleukin-6 biosynthetic process
positive regulation of circadian sleep/wake cycle, non-REM sleep
negative regulation of insulin secretion
decidualization
negative regulation of inflammatory response
cartilage development
positive regulation of corticotropin secretion
positive regulation of cortisol secretion
positive regulation of synapse assembly
excitatory postsynaptic potential
positive regulation of growth hormone secretion
positive regulation of growth hormone receptor signaling pathway
positive regulation of protein tyrosine kinase activity
negative regulation of energy homeostasis
positive regulation of energy homeostasis
G-protein coupled receptor binding
protein binding
growth hormone-releasing hormone activity
protein tyrosine kinase activator activity
ghrelin receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0