Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017528.2 | NP_001017528.1 | 9 | 1430 | ||
NM_001017529.2 | NP_001017529.1 | 8 | 1313 | |||
NM_001017530.1 | NP_001017530.1 | 8 | 1487 | |||
NM_001198721.1 | NP_001185650.1 | 10 | 2143 | |||
NM_015366.3 | NP_056181.2 | 9 | 1604 | |||
NM_181333.3 | NP_851850.1 | 8 | 1465 | |||
GenBank mRNA | AF177331.1 | - | 8 | 1409 | ||
AK000192.1 | - | 4 | 1523 | |||
AK055848.1 | - | 10 | 1865 | |||
AK056787.1 | - | 2 | 1551 | |||
AK091884.1 | - | 9 | 1355 | |||
AK291564.1 | - | 8 | 1455 | |||
BC016921.1 | - | 4 | 1712 | 4413787 |