Gene Symbol: C6orf25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025260.3 | NP_079536.2 | 6 | 693 | ||
NM_138272.2 | NP_612116.1 | 6 | 674 | |||
NM_138273.2 | NP_612117.1 | 5 | 602 | |||
NM_138274.2 | NP_612118.1 | 4 | 561 | |||
NM_138275.2 | NP_612119.1 | 4 | 542 | |||
NM_138277.2 | NP_612121.1 | 5 | 818 | |||
XM_011514920.2 | XP_011513222.1 | 6 | 708 | |||
XM_011514921.2 | XP_011513223.1 | 6 | 727 | |||
XM_017011330.1 | XP_016866819.1 | 4 | 754 | |||
XM_017011331.1 | XP_016866820.1 | 4 | 992 | |||
XM_017011332.1 | XP_016866821.1 | 5 | 636 | |||
GenBank mRNA | AJ292259.1 | - | 6 | 653 | ||
AJ292260.1 | - | 6 | 634 | |||
AJ292261.1 | - | 5 | 562 | |||
AJ292262.1 | - | 4 | 521 | |||
AJ292263.1 | - | 4 | 502 | |||
AJ292264.1 | - | 5 | 771 | |||
AJ292265.1 | - | 5 | 778 | |||
AK092392.1 | - | 6 | 664 |