Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP1M2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300887.1 | NP_001287816.1 | 5 | 571 | ||
NM_005498.4 | NP_005489.2 | 5 | 571 | |||
GenBank mRNA | AF020797.2 | - | 5 | 534 | ||
AK023863.1 | - | 5 | 572 | |||
AK222946.1 | - | 5 | 537 | |||
AK315689.1 | - | 5 | 562 | |||
BC003387.2 | - | 5 | 509 | 3446133 | ||
BC003612.2 | - | 5 | 508 | 2988458 | ||
BC005021.1 | - | 5 | 571 | 3139433 |