Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005500.2 | NP_005491.1 | 7 | 860 | ||
NR_027280.1 | 7 | 909 | ||||
XM_006722963.2 | XP_006723026.1 | 7 | 860 | |||
XM_017026136.1 | XP_016881625.1 | 7 | 731 | |||
GenBank mRNA | AF046025.2 | - | 7 | 797 | ||
AF090385.2 | - | 7 | 805 | |||
AF110956.1 | - | 7 | 752 | |||
AF161489.1 | - | 7 | 808 | |||
AK027595.1 | - | 8 | 982 | |||
AK055481.1 | - | 7 | 825 | |||
AK090645.1 | - | 7 | 878 | |||
AK302287.1 | - | 6 | 535 | |||
AK315624.1 | - | 7 | 802 | |||
BC000344.2 | - | 7 | 813 | 2823028 | ||
BC003611.2 | - | 7 | 781 | 2988252 | ||
BC018271.1 | - | 7 | 820 | 3452340 | ||
BT007290.1 | - | 7 | 752 |