Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145713.1 | NP_001139185.1 | 2, 3 | 311 | ||
NM_001145714.1 | NP_001139186.1 | 2, 3 | 311 | |||
NM_005500.2 | NP_005491.1 | 2, 3 | 311 | |||
NR_027280.1 | 2, 3 | 360 | ||||
XM_006722963.2 | XP_006723026.1 | 2, 3 | 311 | |||
XM_006722965.3 | XP_006723028.1 | 2, 3 | 311 | |||
XM_017026135.1 | XP_016881624.1 | 2, 3 | 311 | |||
XM_017026136.1 | XP_016881625.1 | 2, 3 | 182 | |||
GenBank mRNA | AF046025.2 | - | 2, 3 | 248 | ||
AF090385.2 | - | 2, 3 | 256 | |||
AF110956.1 | - | 2, 3 | 203 | |||
AF161489.1 | - | 2, 3 | 259 | |||
AK021978.1 | - | 2, 3 | 305 | |||
AK027595.1 | - | 2, 3 | 275 | |||
AK055481.1 | - | 2, 3 | 276 | |||
AK090645.1 | - | 2, 3 | 329 | |||
AK315624.1 | - | 2, 3 | 253 | |||
BC000344.2 | - | 2, 3 | 264 | 2823028 | ||
BC003611.2 | - | 2, 3 | 232 | 2988252 | ||
BC018271.1 | - | 2, 3 | 271 | 3452340 | ||
BT007290.1 | - | 2, 3 | 203 | |||
DA047601.1 | - | 2, 3 | 316 | |||
DA086973.1 | - | 2, 3 | 312 | |||
DC297573.1 | - | 2, 3 | 311 | |||
DC312737.1 | - | 2, 3 | 360 |