Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R14A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243947.1 | NP_001230876.1 | 3 | 658 | ||
NM_033256.2 | NP_150281.1 | 4 | 739 | |||
GenBank mRNA | AB056508.1 | - | 4 | 680 | ||
AB056509.1 | - | 3 | 599 | |||
AF123763.1 | - | 4 | 530 | |||
AY050670.1 | - | 4 | 524 | |||
BC021089.1 | - | 4 | 504 | 5019808 |