Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKCDBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145040.2 | NP_659477.2 | 2 | 855 | ||
GenBank mRNA | AF339881.1 | - | 2 | 857 | ||
BC011585.1 | - | 2 | 855 | 4620139 |