Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRGPRE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039165.2 | NP_001034254.2 | 2 | 871 | ||
GenBank mRNA | AK293940.1 | - | 2 | 871 | ||
AY255572.1 | - | 1 | 393 | |||
BC104889.1 | - | 1 | 595 | 8143892 | ||
BC112203.1 | - | 1 | 607 | 8327671 |
Set Membership: |
G-protein coupled receptor |