Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080621.4 | NP_542188.1 | 5 | 1122 | ||
XM_005260199.4 | XP_005260256.1 | 7 | 2259 | |||
XM_006723705.3 | XP_006723768.1 | 6 | 1021 | |||
XM_011528565.2 | XP_011526867.1 | 6 | 1891 | |||
XM_017027671.1 | XP_016883160.1 | 6 | 2139 | |||
GenBank mRNA | BC067362.1 | - | 5 | 1119 | 6672007 |