Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SCAMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320778.1 | NP_001307707.1 | 6 | 701 | ||
NM_005697.4 | NP_005688.2 | 5 | 572 | |||
XM_017021856.1 | XP_016877345.1 | 6 | 626 | |||
GenBank mRNA | AF005038.2 | - | 5 | 477 | ||
AK096376.1 | - | 4 | 463 | |||
AK096794.1 | - | 4 | 2361 | |||
AK291884.1 | - | 5 | 558 | |||
AK315516.1 | - | 5 | 521 | |||
BC001376.2 | - | 5 | 522 | 3050527 | ||
BC004385.1 | - | 5 | 522 | 3617954 | ||
CD300387.1 | - | 6 | 677 | |||
CR456800.1 | - | 5 | 448 |