Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPBAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077191.1 | NP_001070659.1 | 2 | 953 | ||
NM_001077194.1 | NP_001070662.1 | 2 | 445 | |||
NM_001321950.1 | NP_001308879.1 | 2 | 819 | |||
NM_170699.2 | NP_733800.1 | 2 | 332 | |||
XM_011510743.1 | XP_011509045.1 | 2 | 1440 | |||
XM_017003467.1 | XP_016858956.1 | 3 | 1557 | |||
XM_017003468.1 | XP_016858957.1 | 3 | 2178 | |||
XM_017003469.1 | XP_016858958.1 | 3 | 1670 | |||
GenBank mRNA | AB086170.1 | - | 1 | 85 | ||
AB089307.1 | - | 1 | 85 | |||
AK122658.1 | - | 2 | 819 | |||
AK122660.1 | - | 2 | 953 | |||
BC033625.1 | - | 2 | 316 | 5221127 |
Set Membership: |
![]() |