Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XAGE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130776.1 | NP_570132.1 | 5 | 390 | ||
NM_133179.2 | NP_573440.1 | 5 | 421 | |||
XM_011530768.2 | XP_011529070.1 | 5 | 487 | |||
XM_011530769.2 | XP_011529071.1 | 5 | 450 | |||
XM_011530770.2 | XP_011529072.1 | 5 | 414 | |||
GenBank mRNA | AF318372.1 | - | 5 | 402 | ||
AJ318881.1 | - | 5 | 390 | |||
BC062680.1 | - | 5 | 425 | 4619720 |