Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF778 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201407.1 | NP_001188336.1 | 3 | 441 | ||
NM_182531.3 | NP_872337.2 | 3 | 441 | |||
NR_037705.1 | 3 | 441 | ||||
XM_005256288.2 | XP_005256345.1 | 3 | 427 | |||
XM_011522940.2 | XP_011521242.1 | 3 | 394 | |||
XM_017023016.1 | XP_016878505.1 | 3 | 420 | |||
XM_017023017.1 | XP_016878506.1 | 3 | 420 | |||
GenBank mRNA | AK056437.1 | - | 3 | 434 | ||
BC143524.1 | - | 2 | 158 | |||
DA297948.1 | - | 3 | 441 | |||
DC324714.1 | - | 3 | 432 | |||
DR001516.1 | - | 3 | 429 |