Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCE4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178356.3 | NP_848133.1 | 1 | 114 | ||
GenBank mRNA | BC113446.1 | - | 1 | 114 | 8322498 | |
BC113448.1 | - | 1 | 114 | 8322500 |