Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNUPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042581.1 | NP_001036046.1 | 3, 4 | 643 | ||
NM_001042588.1 | NP_001036053.1 | 3, 4 | 448 | |||
NM_005701.3 | NP_005692.1 | 3, 4 | 748 | |||
GenBank mRNA | AF039029.1 | - | 3, 4 | 393 | ||
AK054641.1 | - | 4, 5 | 881 | |||
AK289475.1 | - | 3, 4 | 444 | |||
BC004203.2 | - | 3, 4 | 379 | 3605328 | ||
BU945130.1 | - | 3, 4 | 542 | |||
CD244425.1 | - | 3, 4 | 643 | |||
CR456811.1 | - | 2, 3 | 303 |