Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NRBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178564.3 | NP_848659.2 | 18 | 2545 | ||
XM_017013380.1 | XP_016868869.1 | 17 | 2636 | |||
XM_017013383.1 | XP_016868872.1 | 17 | 2489 | |||
GenBank mRNA | AF318376.1 | - | 8 | 1884 | ||
AK094017.1 | - | 2 | 1530 | |||
AL137662.1 | - | 16 | 2474 | |||
AL834530.1 | - | 11 | 1784 | |||
BC071605.1 | - | 17 | 2708 |