Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005287.1 | NP_001005287.1 | 1 | 315 |
Gene Symbol: OR2A42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001802.2 | NP_001001802.2 | 1 | 315 |