Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM205 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145416.1 | NP_001138888.1 | 3 | 334 | ||
NM_001321112.1 | NP_001308041.1 | 3 | 443 | |||
NM_001321113.1 | NP_001308042.1 | 3 | 253 | |||
NM_001321114.1 | NP_001308043.1 | 3 | 362 | |||
NM_033408.3 | NP_212133.1 | 3 | 453 | |||
NM_198536.2 | NP_940938.1 | 2 | 832 | |||
XM_005259899.2 | XP_005259956.1 | 3 | 539 | |||
XM_017026769.1 | XP_016882258.1 | 1 | 144 | |||
GenBank mRNA | AK127147.1 | - | 2 | 2719 | ||
AK291979.1 | - | 3 | 313 | |||
AY358949.1 | - | 3 | 274 | |||
BC064948.1 | - | 2 | 573 | 6025162 | ||
BC091472.1 | - | 3 | 261 | 30335220 | ||
BE889078.1 | - | 3 | 453 | |||
BM352837.1 | - | 3 | 212 | |||
BM451378.1 | - | 3 | 339 | |||
BQ639667.1 | - | 3 | 370 | |||
CB994930.1 | - | 3 | 288 |