Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR10G4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004462.1 | NP_001004462.1 | 1 | 370 |
Gene Symbol: OR10G9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001953.1 | NP_001001953.1 | 1 | 370 |