Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ARPC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270439.1 | NP_001257368.1 | 1 | 355 | ||
NM_005717.3 | NP_005708.1 | 1 | 355 | |||
GenBank mRNA | AF006088.1 | - | 1 | 97 | ||
AF017807.1 | - | 1 | 76 | |||
AK129598.1 | - | 1 | 140 | |||
BC057237.1 | - | 1 | 175 | 3931697 | ||
BC071857.1 | - | 1 | 276 | 5922521 | ||
BI758106.1 | - | 1 | 264 | |||
CN291383.1 | - | 1 | 355 |