Search Thermo Fisher Scientific
Gene Symbol: PLA2G2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316722.1 | NP_001303651.1 | 2 | 114 | ||
XM_017001270.1 | XP_016856759.1 | 1 | 881 | |||
GenBank mRNA | AW135717.1 | - | 1 | 109 |