Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARPC1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005720.3 | NP_005711.1 | 3 | 215 | ||
XM_006715825.1 | XP_006715888.1 | 3 | 226 | |||
XM_006715826.1 | XP_006715889.1 | 3 | 210 | |||
GenBank mRNA | AF006084.1 | - | 3 | 175 | ||
AK314818.1 | - | 3 | 160 | |||
AU143525.1 | - | 3 | 215 | |||
AY007111.1 | - | 1 | 45 | |||
BC002562.2 | - | 3 | 148 | 3138319 | ||
BC002988.1 | - | 3 | 184 | 2821335 | ||
BC007555.1 | - | 3 | 146 | 2967684 |