Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168412.1 | NP_001161884.1 | 3, 4 | 522 | ||
NM_005724.5 | NP_005715.1 | 4, 5 | 714 | |||
NM_198902.2 | NP_944492.1 | 3, 4 | 639 | |||
XM_017021857.1 | XP_016877346.1 | 4, 5 | 323 | |||
GenBank mRNA | AF054840.1 | - | 4, 5 | 440 | ||
AF133423.1 | - | 4, 5 | 514 | |||
AK001305.1 | - | 4, 5 | 560 | |||
AK001310.1 | - | 4, 5 | 546 | |||
AK001326.1 | - | 4, 5 | 657 | |||
AK027793.1 | - | 1, 2 | 1765 | |||
AK075336.1 | - | 4, 5 | 595 | |||
AK075482.1 | - | 3, 4 | 520 | |||
AK293677.1 | - | 5, 6 | 503 | |||
AK298157.1 | - | 3, 4 | 403 | |||
BC000704.2 | - | 4, 5 | 615 | 3349181 | ||
BC004280.1 | - | 4, 5 | 576 | 3616380 | ||
BC009248.2 | - | 4, 5 | 579 | 2964568 | ||
BC011206.1 | - | 4, 5 | 608 | 3457158 |