Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FAM90A25P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073395.1 | 4 | 1700 | |||
GenBank mRNA | AL832996.1 | - | 4 | 1699 |
Gene Symbol: FAM90A7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046343.1 | 3 | 1645 | |||
GenBank mRNA | AK301810.1 | - | 4 | 1646 |